Learn More
MSC Tingley Jacket, Polyurethane on Nylon, Gold, 2X-Large, 1 ea
Catalog No. 5000793696
Encompass
This exterior coated polyurethane on nylon jacket is lightweight and durable. It offers comfort and flexibility, and resists fats, oils, pine tars, gasoline, grease, and various other chemicals.
  • Batwing sleeve for maximum freedom of movement
  • Therm-O-Rad sealed seams ensure 100% waterproof protection
  • Storm fly front seals out wind, rain, and contaminants
  • Hood snaps for optional hood attachment
  • Heavy-duty snaps
  • Mildew resistant
  • An exterior coating of 2 mil provides snag and abrasion resistance
  • Sleek coating allows for easy decontamination and shedding liquids
Catalog No. 50-007-93696 Supplier MSC Supplier No. 96549282
This exterior coated polyurethane on nylon jacket is lightweight and durable. It offers comfort and flexibility, and resists fats, oils, pine tars, gasoline, grease, and various other chemicals.
  • Batwing sleeve for maximum freedom of movement
  • Therm-O-Rad sealed seams ensure 100% waterproof protection
  • Storm fly front seals out wind, rain, and contaminants
  • Hood snaps for optional hood attachment
  • Heavy-duty snaps
  • Mildew resistant
  • An exterior coating of 2 mil provides snag and abrasion resistance
  • Sleek coating allows for easy decontamination and shedding liquids