The DuraScrim Jacket features a double coating of PVC on polyester, offering durability and high tear strength with an open weave polyester scrim fabric between flame-resistant PVC layers.
- Full cut shoulder with 90° sleeve to body angle for freedom of movement.
- Storm fly front seals out wind, rain, and contaminants.
- Hood snaps compatible with DuraScrim hood (H56107).
- Self-material collar resists contamination.
- Adjustable take-up snaps on sleeves.
- Thicker outside coating for abrasion resistance.
- Lighter interior coating protects against chemical contamination.
- Achieved ASTM F1671 resistance to viral penetration.
- Therm-O-Rad sealed seams provide 100% waterproof and chemical splash protection.
- Flame resistant tested: ASTM D6413 self-extinguishing, <2 sec flameout/afterglow, <6 in char length.