Learn More
MSC Tingley Magnaprene 60 Inch Coat, Flame Resistant, Neoprene, 1 ea
Catalog No. 5000795584
Encompass
This Magnaprene coat is constructed from single-coated neoprene on nylon, providing a durable and protective garment for demanding environments.
  • Olive drab coat, 60 inch
  • Raglan sleeves for maximum freedom of movement
  • Stitched and taped seams provide 100% waterproof protection
  • Storm fly front seals out wind, rain, and contaminants
  • Attached hood and inner cuffs
  • Mildew resistant
  • Nylon fabric for excellent strength and durability
  • Neoprene coating resists abrasion from oils, acids, chemicals, and solvents
  • Flame resistant, tested to ASTM D6413, with self-extinguishing properties
Catalog No. 50-007-95584 Supplier MSC Supplier No. 35116771
This Magnaprene coat is constructed from single-coated neoprene on nylon, providing a durable and protective garment for demanding environments.
  • Olive drab coat, 60 inch
  • Raglan sleeves for maximum freedom of movement
  • Stitched and taped seams provide 100% waterproof protection
  • Storm fly front seals out wind, rain, and contaminants
  • Attached hood and inner cuffs
  • Mildew resistant
  • Nylon fabric for excellent strength and durability
  • Neoprene coating resists abrasion from oils, acids, chemicals, and solvents
  • Flame resistant, tested to ASTM D6413, with self-extinguishing properties