This Magnaprene 48 Inch Coat is a lightweight protective garment made from single-coated neoprene on nylon, designed for demanding industrial environments.
- Features 48 inch length, olive drab color
- Raglan sleeves for maximum freedom of movement
- Stitched and taped seams for 100% waterproof protection
- Storm fly front seals out wind, rain, and contaminants
- Attached hood
- Inner cuffs
- Mildew resistant nylon fabric provides excellent strength and durability
- Neoprene coating for abrasion resistance to oils, acids, chemicals, and solvents
- Flame resistant per ASTM D6413 (self-extinguishing with flameout and afterglow ≤ 2 seconds)
- Resists animal fats, certain acids, alcohols, caustics, oils, and certain solvents
- Ideal for petrochemical, chemical, off shore drilling, mining, fishing, and forestry applications