Learn More
MSC Tingley Magnaprene Overalls, Flame Resistant, Waterproof, Neoprene, 1 ea
Catalog No. 5000788858
Encompass
Lightweight overalls constructed from single-coated neoprene on nylon, offering high-performance durability, flame resistance, and 100% waterproof protection with stitched and taped seams.
  • Stitched and taped seams ensure 100% waterproof protection
  • Take-up snaps on ankle cuffs for adjustable fit
  • Heavy-duty snaps provide maximum performance
  • Mildew resistant material
  • Nylon fabric offers excellent strength and durability
  • Neoprene coating resists oils, acids, chemicals, and solvents
  • Flame resistant per ASTM D6413 (self-extinguishing with flameout and afterglow ≤ 2 seconds)
  • Generous cut allows for maximum freedom of movement
Catalog No. 50-007-88858 Supplier MSC Supplier No. 42596445
Lightweight overalls constructed from single-coated neoprene on nylon, offering high-performance durability, flame resistance, and 100% waterproof protection with stitched and taped seams.
  • Stitched and taped seams ensure 100% waterproof protection
  • Take-up snaps on ankle cuffs for adjustable fit
  • Heavy-duty snaps provide maximum performance
  • Mildew resistant material
  • Nylon fabric offers excellent strength and durability
  • Neoprene coating resists oils, acids, chemicals, and solvents
  • Flame resistant per ASTM D6413 (self-extinguishing with flameout and afterglow ≤ 2 seconds)
  • Generous cut allows for maximum freedom of movement