Learn More
MSC Tingley Magnaprene Overalls, Waterproof, Flame Resistant, Yellow, 1 ea
Catalog No. 5000676639
Encompass
Magnaprene overalls feature single-coated neoprene on nylon, providing a high-performance solution for demanding environments. These lightweight overalls combine durability with resistance to water, flames, and various chemicals.
  • Constructed from neoprene on nylon fabric
  • Plain front, yellow design
  • Ensures 100% waterproof protection with stitched and taped seams
  • Adjustable ankle cuffs with take-up snaps
  • Heavy-duty snaps
  • Nylon fabric ensures excellent strength
  • Neoprene coating resists oils, acids, chemicals, solvents
  • Self-extinguishing flame resistance (ASTM D6413)
  • Mildew resistant
  • Resists animal fats, certain acids, alcohols, caustics
Catalog No. 50-006-76639 Supplier MSC Supplier No. 42596361
Magnaprene overalls feature single-coated neoprene on nylon, providing a high-performance solution for demanding environments. These lightweight overalls combine durability with resistance to water, flames, and various chemicals.
  • Constructed from neoprene on nylon fabric
  • Plain front, yellow design
  • Ensures 100% waterproof protection with stitched and taped seams
  • Adjustable ankle cuffs with take-up snaps
  • Heavy-duty snaps
  • Nylon fabric ensures excellent strength
  • Neoprene coating resists oils, acids, chemicals, solvents
  • Self-extinguishing flame resistance (ASTM D6413)
  • Mildew resistant
  • Resists animal fats, certain acids, alcohols, caustics