Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tingley™ Men's 1200 Storm Overshoes

Catalog No. 19022863 Shop All Tingley Rubber Co. Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Size 10.5 to 12.5 shoes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-022863 Size 10.5 to 12.5 shoes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-022863 Supplier Tingley™ Supplier No. 1200.2X
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Black
For Use With (Equipment) Size 10.5 to 12.5 shoes
Material Rubber
Size 2X-Large
Resistant To Tear
Waterproof Yes
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.