Learn More
MSC Tingley Overalls, Magnaprene, Flame Resistant, Yellow, 3XL, 1 each
Catalog No. 5000766501
Encompass
Magnaprene single coated neoprene on nylon overalls offer classic neoprene performance for demanding environments. These lightweight overalls are designed for petrochemical, chemical, off-shore drilling, mining, fishing, and forestry applications, providing resistance to animal fats, certain acids, alcohols, caustics, oils, and solvents.
  • Neoprene on nylon yellow plain front overalls
  • Stitched and taped seams for 100% waterproof protection
  • Take-up snaps on ankle cuffs for adjustable fit
  • Heavy duty snaps guaranteed not to fail
  • Mildew resistant
  • Neoprene coating provides excellent abrasion performance and resists oils, acids, chemicals, and solvents
  • Nylon fabric provides excellent strength and durability
  • Flame resistant, self-extinguishing with flameout and afterglow ≤2 seconds after removal of ignition source and char length <6 in. (ASTM D6413)
Catalog No. 50-007-66501 Supplier MSC Supplier No. 78305216
Magnaprene single coated neoprene on nylon overalls offer classic neoprene performance for demanding environments. These lightweight overalls are designed for petrochemical, chemical, off-shore drilling, mining, fishing, and forestry applications, providing resistance to animal fats, certain acids, alcohols, caustics, oils, and solvents.
  • Neoprene on nylon yellow plain front overalls
  • Stitched and taped seams for 100% waterproof protection
  • Take-up snaps on ankle cuffs for adjustable fit
  • Heavy duty snaps guaranteed not to fail
  • Mildew resistant
  • Neoprene coating provides excellent abrasion performance and resists oils, acids, chemicals, and solvents
  • Nylon fabric provides excellent strength and durability
  • Flame resistant, self-extinguishing with flameout and afterglow ≤2 seconds after removal of ignition source and char length <6 in. (ASTM D6413)