Learn More
MSC Tingley Overalls, Neoprene on Nylon, Waterproof, Flame Resistant, 1 ea
Catalog No. 5000792658
Encompass
Magnaprene Overalls are constructed from lightweight, single-coated neoprene on nylon, providing a robust option for high-performance use and making them among the toughest overalls available for demanding environments.
  • Single-coated neoprene on nylon material
  • Stitched and taped seams for 100% waterproof protection
  • Take-up snaps on ankle cuffs for adjustable fit
  • Heavy-duty snaps guaranteed not to fail
  • Mildew resistant
  • Nylon fabric provides excellent strength and durability
  • Neoprene coating offers abrasion resistance to oils, acids, chemicals, and solvents
  • Flame resistant, tested to ASTM D6413 standards
  • Ideal for petrochemical, chemical, offshore drilling, mining, fishing, and forestry applications
  • Resistant to animal fats, certain acids, alcohols, caustics, oils, and certain solvents
Catalog No. 50-007-92658 Supplier MSC Supplier No. 91055202
Magnaprene Overalls are constructed from lightweight, single-coated neoprene on nylon, providing a robust option for high-performance use and making them among the toughest overalls available for demanding environments.
  • Single-coated neoprene on nylon material
  • Stitched and taped seams for 100% waterproof protection
  • Take-up snaps on ankle cuffs for adjustable fit
  • Heavy-duty snaps guaranteed not to fail
  • Mildew resistant
  • Nylon fabric provides excellent strength and durability
  • Neoprene coating offers abrasion resistance to oils, acids, chemicals, and solvents
  • Flame resistant, tested to ASTM D6413 standards
  • Ideal for petrochemical, chemical, offshore drilling, mining, fishing, and forestry applications
  • Resistant to animal fats, certain acids, alcohols, caustics, oils, and certain solvents