Learn More
MSC Tingley Rain Jacket, 2X-Large, ASTM D6413, Green, 1 ea
Catalog No. 5000782738
Encompass
This jacket provides comprehensive protection in demanding environments, featuring flame-resistant, chemical-resistant, and waterproof properties. Made from polyester, it is designed for various protective needs and meets the ASTM D6413 standard.
  • Flame-resistant, chemical-resistant, and waterproof
  • Mildew-resistant for enhanced durability
  • Attached hood
  • Secure snaps and zipper closure
  • Constructed from durable polyester material
  • Certified to ASTM D6413 standard
Catalog No. 50-007-82738 Supplier MSC Supplier No. 88138599
This jacket provides comprehensive protection in demanding environments, featuring flame-resistant, chemical-resistant, and waterproof properties. Made from polyester, it is designed for various protective needs and meets the ASTM D6413 standard.
  • Flame-resistant, chemical-resistant, and waterproof
  • Mildew-resistant for enhanced durability
  • Attached hood
  • Secure snaps and zipper closure
  • Constructed from durable polyester material
  • Certified to ASTM D6413 standard