Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Tingley Rain Jacket, 2X-Large, Yellow, Polyester, 1 ea

Catalog No. 5000803125
Encompass

This rain jacket provides waterproof protection with a zipper closure and two pockets, designed for a chest size of 52 to 54 in.

  • Garment style: rain jacket
  • Size: 2X-Large
  • Color: yellow
  • Material: polyester
  • Garment type: waterproof
  • Hood type: attached
  • Chest size: 52 to 54 in.
  • Seam style: heat sealed
  • Wrist style: hook & loop
  • Certification type: non-hazardous protection
  • Material thickness: 0.40 mm
  • Closure type: zipper
  • Number of pockets: 2

Catalog No. 50-008-03125 Supplier MSC Supplier No. 31660004
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This rain jacket provides waterproof protection with a zipper closure and two pockets, designed for a chest size of 52 to 54 in.

  • Garment style: rain jacket
  • Size: 2X-Large
  • Color: yellow
  • Material: polyester
  • Garment type: waterproof
  • Hood type: attached
  • Chest size: 52 to 54 in.
  • Seam style: heat sealed
  • Wrist style: hook & loop
  • Certification type: non-hazardous protection
  • Material thickness: 0.40 mm
  • Closure type: zipper
  • Number of pockets: 2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.