Learn More
MSC Tingley Rain Jacket, 4X-Large, Green, Polyester, Zipper, 1 ea
Catalog No. 5000789098
Encompass
This rain jacket offers comprehensive protection, featuring a polyester construction and a zipper closure. It is designed to be chemical and waterproof, providing flame-resistant properties and meeting ASTM D6413 certification.
  • Designed for 70 inch chest size
  • Green color
  • Made from polyester material
  • Zipper closure type
  • Chemical, waterproof, and flame-resistant/retardant
  • Attached hood
  • Top-stitched and taped seams
  • Mildew-resistant material
  • Open wrist style
  • Material weight of 13 oz.
  • Material thickness of 17 mm
  • Certified under ASTM D6413
Catalog No. 50-007-89098 Supplier MSC Supplier No. 56162068
This rain jacket offers comprehensive protection, featuring a polyester construction and a zipper closure. It is designed to be chemical and waterproof, providing flame-resistant properties and meeting ASTM D6413 certification.
  • Designed for 70 inch chest size
  • Green color
  • Made from polyester material
  • Zipper closure type
  • Chemical, waterproof, and flame-resistant/retardant
  • Attached hood
  • Top-stitched and taped seams
  • Mildew-resistant material
  • Open wrist style
  • Material weight of 13 oz.
  • Material thickness of 17 mm
  • Certified under ASTM D6413