Learn More
MSC Tingley Rain Jacket, High-Visibility, Flame Resistant, 2XL, 1 ea
Catalog No. 5000673709
Encompass
This Comfort-Brite Jacket is a high-visibility, flame-resistant rain jacket providing waterproof protection and enhanced safety features for demanding work environments.
  • ANSI/ISEA 107 Type R Class 3 compliant for high visibility
  • Fluorescent background material for excellent daytime visibility
  • Silver reflective tape, 2 in. wide, for 360° nighttime conspicuity
  • Therm-O-Rad sealed seams for 100% waterproof protection
  • Zipper/storm fly front seals out wind, rain, and contaminants
  • Non-conductive hardware
  • D-ring access for fall protection harness
  • Flame resistant tested to ASTM D6413, self-extinguishing
  • Mildew resistant and resists many acids, oils, alcohols, salts, and alkalis
Catalog No. 50-006-73709 Supplier MSC Supplier No. 43492305
This Comfort-Brite Jacket is a high-visibility, flame-resistant rain jacket providing waterproof protection and enhanced safety features for demanding work environments.
  • ANSI/ISEA 107 Type R Class 3 compliant for high visibility
  • Fluorescent background material for excellent daytime visibility
  • Silver reflective tape, 2 in. wide, for 360° nighttime conspicuity
  • Therm-O-Rad sealed seams for 100% waterproof protection
  • Zipper/storm fly front seals out wind, rain, and contaminants
  • Non-conductive hardware
  • D-ring access for fall protection harness
  • Flame resistant tested to ASTM D6413, self-extinguishing
  • Mildew resistant and resists many acids, oils, alcohols, salts, and alkalis