Learn More
MSC Tingley Rain Overalls, Size Small, Black, Polyester, 1 ea
Catalog No. 5000795280
Encompass
These rain overalls are designed for general purpose use, crafted from black polyester material with a thickness of 0.3 mm and a weight of 5.5 oz. They offer waterproof protection, heavy-duty suspenders, and take-up snaps, making them suitable for various tasks requiring durable and protective workwear.
  • Waterproof design
  • Heavy-duty suspenders
  • Take-up snaps
  • Large chest pocket
  • Ankle with hook & loop closure and zipper
  • Zippered leg openings
  • Breathable material
  • Hook & loop take-up straps
  • Top-stitched and taped seams
  • Waist size 32 to 34 in.
  • Inseam 28 in.
  • Snap closure type
  • Snap fly type
  • Rollaway hood
  • Certified for non-hazardous protection
Catalog No. 50-007-95280 Supplier MSC Supplier No. 31659790
These rain overalls are designed for general purpose use, crafted from black polyester material with a thickness of 0.3 mm and a weight of 5.5 oz. They offer waterproof protection, heavy-duty suspenders, and take-up snaps, making them suitable for various tasks requiring durable and protective workwear.
  • Waterproof design
  • Heavy-duty suspenders
  • Take-up snaps
  • Large chest pocket
  • Ankle with hook & loop closure and zipper
  • Zippered leg openings
  • Breathable material
  • Hook & loop take-up straps
  • Top-stitched and taped seams
  • Waist size 32 to 34 in.
  • Inseam 28 in.
  • Snap closure type
  • Snap fly type
  • Rollaway hood
  • Certified for non-hazardous protection