Learn More
MSC Tingley Rain Suit, .35mm PVC on Polyester, Medium, 1 ea
Catalog No. 5000781868
Encompass
This 3-piece rain suit provides 100% waterproof protection, designed with a generous cut for ease of movement and comfort. It is a versatile solution for general-purpose applications, offering durability at an economical price.
  • Provides 100% waterproof protection
  • Generously cut for ease of movement and comfort
  • Electronically welded seams
  • Detachable hood
  • Take-up snaps on sleeves and ankle cuffs for adjustable fit
  • Storm fly zipper front jacket
  • Snap fly front overalls
  • Two pockets
Catalog No. 50-007-81868 Supplier MSC Supplier No. 31659865
This 3-piece rain suit provides 100% waterproof protection, designed with a generous cut for ease of movement and comfort. It is a versatile solution for general-purpose applications, offering durability at an economical price.
  • Provides 100% waterproof protection
  • Generously cut for ease of movement and comfort
  • Electronically welded seams
  • Detachable hood
  • Take-up snaps on sleeves and ankle cuffs for adjustable fit
  • Storm fly zipper front jacket
  • Snap fly front overalls
  • Two pockets