Learn More
MSC Tingley Rain Vest, ANSI 107, Reversible, 1 ea
Catalog No. 5000796307
Encompass
This reversible rain vest is designed for safety and durability, offering compliance with ANSI 107 standards. Made from polyester, it features fluorescent lime and black colors, providing high visibility in various conditions.
  • Offers 360° nighttime conspicuity
  • Tapered back cut for added protection
  • Zipper front closure
  • Waterproof
  • Reversible design
Catalog No. 50-007-96307 Supplier MSC Supplier No. 31660095
This reversible rain vest is designed for safety and durability, offering compliance with ANSI 107 standards. Made from polyester, it features fluorescent lime and black colors, providing high visibility in various conditions.
  • Offers 360° nighttime conspicuity
  • Tapered back cut for added protection
  • Zipper front closure
  • Waterproof
  • Reversible design