Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tingley™ Rubber Work High-top Galoshes

Catalog No. 191305822 Shop All Tingley Rubber Co. Products
Encompass_Preferred
Change view
Click to view available options
Size:
2X-Large, 12.5 to 14
3X-Large, 14.5 to 15
Large, 9.5 to 11
Medium, 8 to 9.5
Small, 6 to 7.5
X-Large, 11 to 12.5
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-5822 3X-Large, 14.5 to 15
19-048751 Medium, 8 to 9.5
19-048753 Large, 9.5 to 11
19-051964 X-Large, 11 to 12.5
19-051960 2X-Large, 12.5 to 14
19-051961 Small, 6 to 7.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-130-5822 Supplier Tingley™ Supplier No. 1300.3X
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Won't crack or stiffen, even in cold weather

  • Made from an exclusive stretch-rubber compound
  • Protect against alcohol, and many organic acids, salts and alkalis
  • No hardware to tear out or conduct electricity
  • No lining for easy decontamination
  • Deep, slip-resistant treads
  • Black

Specifications

Closure Type Button
Color Black
For Use With (Application) Food processing, transportation, utilities, construction, general industry, municipalities, and agriculture
Material Rubber
Size 3X-Large, 14.5 to 15
Sole Features Deep-cleated outsole
Style High top galoshes
Resistant To Chemicals, acids, alcohols
Waterproof Yes
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.