Learn More
MSC Tingley Safetyflex Jacket, PVC, Chemical/Flame Resistant, Green, 1 ea
Catalog No. 5000802723
Encompass
The Safetyflex jacket is made from flame-resistant specialty PVC on polyester, offering robust protection and comfort. It resists industrial acids, chemicals, and caustics, ideal for demanding environments.
  • Shoulder cut with 90° sleeve-to-body angle for freedom of movement
  • Therm-O-Rad sealed seams provide 100% liquidproof protection
  • Storm fly front seals out wind, rain, and contaminants
  • Non-conductive, non-corrosive snaps; hidden hardware
  • Collar throat guard with hook & loop closure for splash protection
  • Mildew resistant with soft, supple feel
  • Heavy exterior coating for excellent permeation performance
  • Tapered sleeves accommodate gloves
  • Safety green color for identification
  • Flame resistant (ASTM D6413)
  • Viral penetration resistant (ASTM F1671)
  • Resists industrial acids, chemicals, and caustics
Catalog No. 50-008-02723 Supplier MSC Supplier No. 66545401
The Safetyflex jacket is made from flame-resistant specialty PVC on polyester, offering robust protection and comfort. It resists industrial acids, chemicals, and caustics, ideal for demanding environments.
  • Shoulder cut with 90° sleeve-to-body angle for freedom of movement
  • Therm-O-Rad sealed seams provide 100% liquidproof protection
  • Storm fly front seals out wind, rain, and contaminants
  • Non-conductive, non-corrosive snaps; hidden hardware
  • Collar throat guard with hook & loop closure for splash protection
  • Mildew resistant with soft, supple feel
  • Heavy exterior coating for excellent permeation performance
  • Tapered sleeves accommodate gloves
  • Safety green color for identification
  • Flame resistant (ASTM D6413)
  • Viral penetration resistant (ASTM F1671)
  • Resists industrial acids, chemicals, and caustics