Learn More
MSC Tingley Shoe Cover, Latex, Water-Resistant, Size 7-9, 100/Pk.
Catalog No. 5000770054
Encompass
This Tingley shoe cover is designed for high-speed applications, offering protection and durability. Made from latex, it is water-resistant and suitable for a range of shoe sizes.
  • Elastic closure
  • Stands 6 in high
  • ASTM F1671 standard
  • Provides 100% liquid proof
  • Textured outsole
  • Water-resistant
  • Fits shoe size 7 to 9
Catalog No. 50-007-70054 Supplier MSC Supplier No. 42954073
This Tingley shoe cover is designed for high-speed applications, offering protection and durability. Made from latex, it is water-resistant and suitable for a range of shoe sizes.
  • Elastic closure
  • Stands 6 in high
  • ASTM F1671 standard
  • Provides 100% liquid proof
  • Textured outsole
  • Water-resistant
  • Fits shoe size 7 to 9