Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tingley™ Snugleg Boots: Triple Dipped, Steel Toe

Catalog No. 19960015 Shop All Tingley Rubber Co. Products
Encompass_Preferred
Change view
Click to view available options
Size:
5
6
7
8
9
10
11
12
13
14
15
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-960-015 15
19-960-005 5
19-960-006 6
19-960-009 9
19-960-011 11
19-960-007 7
19-960-008 8
19-960-010 10
19-960-012 12
19-960-013 13
19-960-014 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 19-960-015 Supplier Tingley™ Supplier No. MB922B.15
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Every boot is dipped with a neoprene coating and sealant, so the foot is surrounded by a layer of 100% waterproof and chemical-resistant neoprene for protection and comfort

  • Chevron or Safety-Loc outsoles offer abrasion resistance and provide excellent slip performance on wet-contaminated or wet-clean surfaces
  • Hand-layered construction allows for a soft flexible upper and more material in the toe and heel contact points, for optimum comfort and protection
  • Neoprene rubber will outwear standard rubber boots in solvents, caustics, hydrocarbons, oils, fats and acids
  • Removable polyurethane contour cushion insoles for maximum comfort
  • Snugleg last design hugs the foot for all day working comfort
  • Dense fabric liner for easy on and off; also wicks moisture and condensation
  • Color: brick red upper with brown sole
  • Steel shank for added arch support

Specifications

Certifications/Compliance ASTM F2413 M/I/75/C/75
Color Red, Brown
For Use With (Application) Designed for tough industrial environments
Height (English) 12.5 in.
Liner Material Fabric
Material Neoprene
Shank Material Steel
Size 15
Sole Features Chevron outsole, Polyurethane contour cushion insole
Toe Material Steel
WARNING: Cancer - www.P65Warnings.ca.gov

Chemical Resistance: Animal fats and blood, oils, certain acids, alcohols, alkalies, and certain solvents. Electrical Hazard soles and heels are intended to reduce the hazards due to accidental contact with live electrical circuits, electrically energized conductors, parts or apparatus. Electric Hazard soles and heels are not intended for wear in those work environments where volatile chemicals or explosives may be present, where conductive footwear is required.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.