Learn More
MSC Tingley Work Boot, Composite Toe, Knee, Size 12, 1 pair
Catalog No. 5000769215
Encompass
The Tingley Flite Safety Toe Knee Boot is constructed from Aerex 1.5.5®, a proprietary, chemical-resistant, thermally-insulative microcellular polymer. This makes the boot over 40% lighter than traditional PVC and rubber boots. It features a lightweight upper, cushioned heel, slip-resistant nitrile rubber outsole, and composite toe for enhanced comfort and safety in food processing and petrochemical environments.
  • Constructed from Aerex 1.5.5® microcellular polymer for over 40% lighter weight.
  • Composite safety toe meets ASTM F2413 M/I/C EH*, non-metallic.
  • Seamless upper is 100% liquid proof.
  • Cleated, slip-resistant nitrile rubber outsole.
  • Insulated with trapped air bubbles.
  • Anti-fatigue design reduces back and leg strain.
  • Calf-relief topline for easier on and off.
  • Tread wear indicators included.
Catalog No. 50-007-69215 Supplier MSC Supplier No. 45996386
The Tingley Flite Safety Toe Knee Boot is constructed from Aerex 1.5.5®, a proprietary, chemical-resistant, thermally-insulative microcellular polymer. This makes the boot over 40% lighter than traditional PVC and rubber boots. It features a lightweight upper, cushioned heel, slip-resistant nitrile rubber outsole, and composite toe for enhanced comfort and safety in food processing and petrochemical environments.
  • Constructed from Aerex 1.5.5® microcellular polymer for over 40% lighter weight.
  • Composite safety toe meets ASTM F2413 M/I/C EH*, non-metallic.
  • Seamless upper is 100% liquid proof.
  • Cleated, slip-resistant nitrile rubber outsole.
  • Insulated with trapped air bubbles.
  • Anti-fatigue design reduces back and leg strain.
  • Calf-relief topline for easier on and off.
  • Tread wear indicators included.