Learn More
Tingley™ Workbrutes™ PVC Knee-High Overshoes
Catalog No. 19999342A Shop All Tingley Rubber Co. Products
Change view
Click to view available options
Size:
Large, 11 to 13
Small, 6.5 to 8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-999-342A Large, 11 to 13
19-999-342C Small, 6.5 to 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-999-342A Supplier Tingley™ Supplier No. 35141.LG
Add to Cart
Add to Cart
This item is not returnable. View return policy
These PVC overshoes have thicker and deeper cleats for a better grip and longer wear
  • Injection-molded, seamless construction for 100% waterproof construction
  • Injection molding puts less material in the upper, eliminating unnecessary weight, for reduced leg fatigue
  • More material is added to the sole at the heel, toe and cleats for longer life
  • Thicker tread has deeper cleats with sharper edges for more traction
  • Key interior entry points are specially textured to enable overshoes to easily slip on and off over rubber sole shoes
  • No-pinch molded-in button and two button holes provide a snug fit and protection
  • Resistant to fats, certain acids, hydrocarbons and caustics
Food processing, construction, general industry, transportation and hazardous waste cleanup
Specifications
Color Black
Height (English) 14 in.
Material PVC
Size Large, 11 to 13
WARNING: Cancer - www.P65Warnings.ca.gov
Not recommended for use in ketones, aldehydes and many solvents.