Learn More
Medchemexpress LLC HY-B0177 100mg Medchemexpress, Tinidazole CAS:19387-91-8 Purity:>98%
SDP
Catalog No. 502082396
Encompass_Preferred
Medchemexpress, HY-B0177 100mg Tinidazole CAS:19387-91-8 Tinidazole, an orally available antibacterial agent, is a 5-nitroimidazole with selective activity against anaerobic bacteria and protozoa. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2396 Supplier Medchemexpress LLC Supplier No. HYB0177100MG
Medchemexpress, HY-B0177 100mg Tinidazole CAS:19387-91-8 Tinidazole, an orally available antibacterial agent, is a 5-nitroimidazole with selective activity against anaerobic bacteria and protozoa. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.