Learn More
Sigma Aldrich Fine Chemicals Biosciences TinII chloride dihydrate A25KG
Catalog No. 502906996
Encompass
Crystals of SnCl2.2H2O exhibit exhibit monoclinic crystal system and space group P21/c.
Catalog No. 50-290-6996 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2435232.5KG
Crystals of SnCl2.2H2O exhibit exhibit monoclinic crystal system and space group P21/c.