Learn More
Sigma Aldrich Fine Chemicals Biosciences TinII chloride dihydrate p500G
Catalog No. 502897745
Encompass
Tin(II) chloride dihydrate is the hydrated salt of tin(II). Its crystals are monoclinic and their structure has been investigated by single crystal X-ray diffraction method. Cell parameters were reported to be a9.313 b7.250 c8.970A… 11455AE and Z4.
Catalog No. 50-289-7745 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 31669500G
Tin(II) chloride dihydrate is the hydrated salt of tin(II). Its crystals are monoclinic and their structure has been investigated by single crystal X-ray diffraction method. Cell parameters were reported to be a9.313 b7.250 c8.970A… 11455AE and Z4.