Learn More
Grainger Channellock Tinner Snip, Straight, Molybdenum Alloy Steel, 8.3 in., 1 ea
Catalog No. 502758281
Encompass_Preferred
These 8-inch tinner snips are designed for precision cutting of sheet metal, vinyl, plastic, rubber, and canvas. Engineered for versatility, they effectively perform straight and wide curve cuts in any direction.
  • Forged from molybdenum alloy steel for strength and durability
  • Precision-formed blades ensure complete cutting strokes and accurate trim cuts
  • 24-gauge cold-rolled steel cutting capacity
  • Comfortable blue grips for extended use
  • Overall length of 8.3 in. with 1.75 in. cutting edge
  • Made in the USA
Catalog No. 50-275-8281 Supplier Grainger Supplier No. 150F39
These 8-inch tinner snips are designed for precision cutting of sheet metal, vinyl, plastic, rubber, and canvas. Engineered for versatility, they effectively perform straight and wide curve cuts in any direction.
  • Forged from molybdenum alloy steel for strength and durability
  • Precision-formed blades ensure complete cutting strokes and accurate trim cuts
  • 24-gauge cold-rolled steel cutting capacity
  • Comfortable blue grips for extended use
  • Overall length of 8.3 in. with 1.75 in. cutting edge
  • Made in the USA