Learn More
Medchemexpress LLC Tinoridine (hydrochloride) | 25913-34-2 | 100.0% | 352.88 | 25 MG
SDP
Catalog No. 5000346002
Encompass_Preferred
Tinoridine hydrochloride is a nonsteroidal anti-inflammatory agent designed for research use. It demonstrates potent radical scavenger and antiperoxidative activity, reducing stable free radicals and inhibiting lipid peroxidation in rat liver microsomes. In vivo studies indicate it can significantly reduce carbon tetrachloride-induced alterations in liver enzyme activities, promoting rapid recovery.
  • Potent radical scavenger and antiperoxidative activity
  • Reduces stable free radicals
  • Inhibits lipid peroxidation in rat liver microsomes
  • Promotes rapid recovery from CCl4-induced liver enzyme alterations
  • Intended for laboratory chemical use
Catalog No. 50-003-46002 Supplier Medchemexpress LLC Supplier No. 25MGHY111354
Tinoridine hydrochloride is a nonsteroidal anti-inflammatory agent designed for research use. It demonstrates potent radical scavenger and antiperoxidative activity, reducing stable free radicals and inhibiting lipid peroxidation in rat liver microsomes. In vivo studies indicate it can significantly reduce carbon tetrachloride-induced alterations in liver enzyme activities, promoting rapid recovery.
  • Potent radical scavenger and antiperoxidative activity
  • Reduces stable free radicals
  • Inhibits lipid peroxidation in rat liver microsomes
  • Promotes rapid recovery from CCl4-induced liver enzyme alterations
  • Intended for laboratory chemical use