Learn More
Medchemexpress LLC Tinoridine hydrochloride | 25913-34-2 | 99.9% | 352.88 | 10 MG
SDP
Catalog No. 5000343428
Encompass_Preferred
Tinoridine hydrochloride is a nonsteroidal anti-inflammatory agent that also exhibits potent radical scavenger and antiperoxidative activity. It reduces stable free radicals, inhibits lipid peroxidation, and is oxidized during lipid peroxidation. In vivo, it significantly reduces CCl4-induced alterations in enzyme activities, promoting rapid recovery.
  • Nonsteroidal anti-inflammatory agent
  • Potent radical scavenger
  • Antiperoxidative activity
Catalog No. 50-003-43428 Supplier Medchemexpress LLC Supplier No. HY11135410MM/1ML
Tinoridine hydrochloride is a nonsteroidal anti-inflammatory agent that also exhibits potent radical scavenger and antiperoxidative activity. It reduces stable free radicals, inhibits lipid peroxidation, and is oxidized during lipid peroxidation. In vivo, it significantly reduces CCl4-induced alterations in enzyme activities, promoting rapid recovery.
  • Nonsteroidal anti-inflammatory agent
  • Potent radical scavenger
  • Antiperoxidative activity