Learn More
Medchemexpress LLC Tinoridine hydrochloride | 25913-34-2 | 99.95% | 352.88 | 50 MG
SDP
Catalog No. 5000343151
Encompass_Preferred
Tinoridine hydrochloride is a nonsteroidal anti-inflammatory agent that also has potent radical scavenger and antiperoxidative activity. It is intended for research use only.
  • Reduces stable free radicals and inhibits lipid peroxidation.
  • Oxidized during lipid peroxidation.
  • Does not affect the reduction of cytochrome c caused by superoxide radical.
  • Substantially reduces CCl4-induced alterations in liver microsomal cytochrome P-450 and G6Pase concentrations.
  • Promotes rapid recovery to normal values in hepatic endoplasmic reticulum function after CCl4 administration.
Catalog No. 50-003-43151 Supplier Medchemexpress LLC Supplier No. HY111354100MG
Tinoridine hydrochloride is a nonsteroidal anti-inflammatory agent that also has potent radical scavenger and antiperoxidative activity. It is intended for research use only.
  • Reduces stable free radicals and inhibits lipid peroxidation.
  • Oxidized during lipid peroxidation.
  • Does not affect the reduction of cytochrome c caused by superoxide radical.
  • Substantially reduces CCl4-induced alterations in liver microsomal cytochrome P-450 and G6Pase concentrations.
  • Promotes rapid recovery to normal values in hepatic endoplasmic reticulum function after CCl4 administration.