Learn More
Cayman Chemical TInptIn MetabolIte MC5 10mg
SDP
Catalog No. 502909517
Encompass_Preferred
An active metabolite of tianeptine; selectively induces G protein activation in BRET assays using HEK293T cells expressing human MOR over human DOR (EC50s = 0.454 and >100 µM, respectively); decreases immobility in the forced swim test in wild-type mice, but not MOR knockout mice, indicating MOR-dependent antidepressant-like activity at 30 mg/kg
Catalog No. 50-290-9517 Supplier Cayman Chemical Supplier No. 3669410 MG
An active metabolite of tianeptine; selectively induces G protein activation in BRET assays using HEK293T cells expressing human MOR over human DOR (EC50s = 0.454 and >100 µM, respectively); decreases immobility in the forced swim test in wild-type mice, but not MOR knockout mice, indicating MOR-dependent antidepressant-like activity at 30 mg/kg