Learn More
3M™ Tinted Lens Cover
Catalog No. 19813734 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
3M™ Full Facepiece 7000 Series
Catalog No. For Use With (Equipment)
19-813-734 3M™ Full Facepiece 7000 Series
1 options
This item is not returnable. View return policy
Catalog No. 19-813-734 Supplier 3M™ Supplier No. 7000126281
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use with 7000 series full facepieces. Not for use in welding or other applications that require reduction of UV or IR radiation beyond the capability of normal nonprescription sunglasses.
  • Accessory to shield eyes from bright, intense light (not for welding)
  • Tinting relaxes eyes to make it easier to focus on outdoor work
  • Designed for use with 3M Full Facepiece 7000 Series
  • Cover fits easily over lens
  • Approved 3M protection accessory
TRUSTED_SUSTAINABILITY
Specifications
Material Polyester Film
Product Type Replacement Parts and Accessories
For Use With (Application) Assembly and Mechanical
For Use With (Equipment) 3M™ Full Facepiece 7000 Series
Model 7986
Quantity 25/Bag