Description
For use with Nitrate LR Checkit Comparator Kit.
- The Lovibond™ Checkit™ comparator is a compact, lightweight colorimetric unit
- The D55 Checkit Comparator contains mirror optics which extend the cell path length to 55mm, allowing greater accuracy at lower concentrations
- Suitable for both mobile and static water analysis
- Available with a number of different color scales
- Provides the basis for a comprehensive easy to to to use analysis system
- Each Checkit™ Comparator Disc contains a continuous color scale for exact color matching between standard and sample
Specifications
Specifications
| For Use With (Equipment) | Nitrate LR CHECKIT Comparator Kit |
| Type | CHECKIT™ Comparator Nitrate Test Tube |