Description
- The Lovibond™ Checkit™ comparator is a compact, lightweight colorimetric unit
- Suitable for both mobile and static water analysis
- Available with a number of different color scales
- Provides the basis for a comprehensive easy to to to use analysis system
- Each Checkit™ Comparator Disc contains a continuous colour scale for exact color matching between standard and sample
- Each Checkit™ Comparator Kit is supplied in a carry case with comparator, vials, discs, reagents and method instructions for all tests
Specifications
Specifications
| Description | Dia.: 13.5mm |
| For Use With (Application) | Water Quality Testing |
| For Use With (Equipment) | Checkit Comprator Tests |
| Includes | Carry case with comparator, vials, discs, reagents and method instructions for all tests |
| Type | Checkit Comprator with Standard Optics |