Learn More
Lovibond™ Nitrification Inhibitor Set-Up
Catalog No. 15398525 Shop All Tintometer Group Products
Change view
Click to view available options
Supplied with dispenser and 35g reagent vial
Catalog No.
15-398-525 Supplied with dispenser and 35g reagent vial
1 options
Catalog No. 15-398-525 Supplier Lovibond™ Supplier No. 194920
Add to Cart
Add to Cart
Supplied with Dispenser and 35g of reagent
  • For BOD analysis
Specifications
Includes Dispenser and 35g reagent vial
Type Nitrification Inhibitor Set-Up