Learn More
Lovibond™ Tintometer™ Phosphate, total LR Tube Test, 0.07 to 3mg/L and 0.2 to 10mg/L
Shop All Tintometer Group ProductsDescription
- Parameter: Total Phosphate
- Method: Phosphomolybdic acid
- For use in test kits, colorimeters and spectrophotometers
- Case of 24
- Includes:
- Reaction Tube, LR
- Phosphate-101
- 2 Calibration Tubes
- Phosphate-102 Solution
- Phosphate-103 Reagent
Specifications
Specifications
| For Use With (Equipment) | Test Kits, Colorimeters and Spectrophotometers |
| Includes | Reaction Tube, LR, Phosphate-101, Calibration Tube - 2 ct., Phosphate-102 Solution, Phosphate-103 Reagent |
| No. of Tests | 24 |
| Range | 0.07 to 3 mg/L and 0.2 to 10 mg/L |
| Test Method | Phosphomolybdic acid |
| Type | Phosphate, Total LR Tube Test |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.