Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Tintometer™ Potassium Sodium Tartrate Test, 100mL

Catalog No. 09050146 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-050-146 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-050-146 Supplier Lovibond™ Supplier No. 465201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Lovibond test kits, colorimeters and spectrophotometers

  • Potassium Sodium Tartrate test for sea water, 100mL
  • As a rule, liquid reagents do not consist of a single preparation but comprise several components that need to be added to the sample in a certain order. As both the size and the number of drops have a decisive effect on the resultant color complex, the reagents need to be added with a high degree of precision. The useful life of liquid reagents is reduced by temporary contact with oxygen in the air when the bottle is opened as well as by unsuitable storage environments (presence of sunlight or high temperatures).

Specifications

For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 100 mL
Type Potassium Sodium Tartrate for sea water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.