Learn More
Lovibond™ Tintometer™ Standard Solution Ammonia, 1.3 mg/L NH4 = 1.0mg/L N
Shop All Tintometer Group ProductsDescription
- Standard Solution Ammonia
- 1.3 mg/L NH4 = 1.0mg/L N
- For use with Lovibond test kits, colorimeters and spectrophotometers
Specifications
Specifications
| Description | 1.3 mg/L NH4 = 1.0mg/L N |
| For Use With (Equipment) | Test Kits, Colorimeters and Spectrophotometers |
| Type | Amonia Solution |