Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Tintometer™ VARIO™ Hydrazine Reagent Solution, 100mL

Catalog No. 09053320 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-053-320 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-053-320 Supplier Lovibond™ Supplier No. 531200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formulated for use in Lovibond test kits, colorimeters and spectrophotometers.

  • Range: 0.01 to 0.6mg/L
  • Parameter: Hydrazine
  • Method: Dimethylamino-benzaldehyde
  • For use in test kits, colorimeters and spectrophotometers
  • Sold individually

Specifications

For Use With (Application) Water Testing
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 100 mL
Test Method Dimethylamino-benzaldehyde
Type VARIO™ Hydrazine Reagent Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.