Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Tintometer™ VARIO™ Iron Ferro Reagent Powder, 25mL

Catalog No. 09050127 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-050-127 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-050-127 Supplier Lovibond™ Supplier No. 530573
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formulated for use in Lovibond test kits, colorimeters and spectrophotometers. Newly redesigned foil packs are easy to handle, easy to open and pour

  • For use in our test kits, colorimeters and spectrophotometers
  • Pre-measured, unit dose reagents
  • Easy to handle, easy to open and easy to pour
  • Case of 100
  • Fast and easy use of VARIO Powder Packs has made them extremely popular for water testing applications in many countries throughout the world
  • Provides more experienced users with a real alternative to existing measurement systems
  • Produced to the same high quality standards that have made Tintometer’s tablet reagents so successful for several decades
  • Parameters from aluminum and chlorine through to sulphate are just some of the well-known tests that are included in the VARIO Powder Pack range

Specifications

For Use With (Application) Water Testing
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
No. of Tests Case of 1000
Quantity 25 mL
Type VARIO™ Iron Ferro Reagent Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.