Learn More
Lovibond™ Tintometer™ VARIO™ Manganese HR Powder Pack Reagent Set, 10mL
Shop All Tintometer Group ProductsDescription
- Range: 0 to 20mg/L
- For use in our test kits, colorimeters and spectrophotometers
- Pre-measured, unit dose reagents
- Easy to handle, easy to open and easy to pour
- Powder pack reagents have a 5 year shelf life
- Case of 100
Specifications
Specifications
| Format | Powdered Reagent |
| For Use With (Application) | Water Testing |
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| No. of Tests | Case of 100 |
| Quantity | 10 mL |
| Range | 0 to 20mg/L |
| Type | Manganese HR Powder Pack Reagent Set |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.