Learn More
Apexbio Technology LLC Tioconazole 65899-73-2 5g
SDP
Catalog No. 502240304
Encompass_Preferred
Tioconazole (65899-73-2) is a small-molecule inhibitor targeting the fungal cytochrome P450-dependent enzyme lanosterol 14 -demethylase It is designed to inhibit ergosterol biosynthesis thereby disrupting fungal cell membrane stability and cellular integrity Tioconazole exerts its biological activity primarily through interference with ergosterol biosynthesis within fungal cell membranes In in vitro studies Tioconazole demonstrates inhibitory activity with MIC50 values below 3 12 mg/L for dermatophyte strains and below 9 mg/L for yeast species Based on these pharmacological properties Tioconazole holds research potential in the study of antifungal mechanisms and in vitro antifungal screening assays
This item is not returnable. View return policy
Catalog No. 50-224-0304 Supplier Apexbio Technology LLC Supplier No. B20515000
Tioconazole (65899-73-2) is a small-molecule inhibitor targeting the fungal cytochrome P450-dependent enzyme lanosterol 14 -demethylase It is designed to inhibit ergosterol biosynthesis thereby disrupting fungal cell membrane stability and cellular integrity Tioconazole exerts its biological activity primarily through interference with ergosterol biosynthesis within fungal cell membranes In in vitro studies Tioconazole demonstrates inhibitory activity with MIC50 values below 3 12 mg/L for dermatophyte strains and below 9 mg/L for yeast species Based on these pharmacological properties Tioconazole holds research potential in the study of antifungal mechanisms and in vitro antifungal screening assays