Learn More
Medchemexpress LLC Tiopronin-d3 | 1189700-74-0 | 99.4% | 1 MG
SDP
Catalog No. 5000407317
Encompass_Preferred
Tiopronin-d3 is the deuterium labeled Tiopronin. Tiopronin acts as a diffusible antioxidant, an antidote to heavy metal poisoning, and a radioprotective agent. It can regulate the rate of cystine precipitation and excretion, showing potential for treating cystinuria, rheumatoid arthritis, and hepatic disorders. For research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-07317 Supplier Medchemexpress LLC Supplier No. HYB0373S11MG
Add to Cart
Add to Cart
Tiopronin-d3 is the deuterium labeled Tiopronin. Tiopronin acts as a diffusible antioxidant, an antidote to heavy metal poisoning, and a radioprotective agent. It can regulate the rate of cystine precipitation and excretion, showing potential for treating cystinuria, rheumatoid arthritis, and hepatic disorders. For research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.