Learn More
Cayman Chemical Tiopronin-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1189700-74-0, 1 mg
SDP
Catalog No. 502947167
Encompass_Preferred
An internal standard for the quantification of tiopronin by GC- or LC-MS
Catalog No. 50-294-7167 Supplier Cayman Chemical Supplier No. 333761 MG
An internal standard for the quantification of tiopronin by GC- or LC-MS