Learn More
Medchemexpress LLC HY-17360 10mg Medchemexpress, Tiotropium (Bromide) CAS:136310-93-5 Purity:>98%
SDP
Catalog No. 501969985
Encompass_Preferred
Medchemexpress, HY-17360 10mg Tiotropium (Bromide) CAS:136310-93-5 Tiotropium Bromide (BA679 BR) is a muscarinic acetylcholine receptor (mAChR) antagonist that blocks the binding of the acetylcholine ligand and subsequent opening of the ligand-gated ion channel. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9985 Supplier Medchemexpress LLC Supplier No. HY1736010MG
Medchemexpress, HY-17360 10mg Tiotropium (Bromide) CAS:136310-93-5 Tiotropium Bromide (BA679 BR) is a muscarinic acetylcholine receptor (mAChR) antagonist that blocks the binding of the acetylcholine ligand and subsequent opening of the ligand-gated ion channel. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.