Learn More
Medchemexpress LLC Tiotropium bromide | 136310-93-5 | 472.42 | 1 ML
SDP
Catalog No. 5000385179
Encompass_Preferred
Tiotropium bromide is a muscarinic acetylcholine receptor (mAChR) antagonist that blocks the binding of the acetylcholine ligand and subsequent opening of the ligand-gated ion channel. It is a 24-hour, anticholinergic bronchodilator used in the management of chronic obstructive pulmonary disease. It improves symptoms of chronic obstructive pulmonary disease complicated by chronic heart failure.
  • Targets mAChR
  • Involved in GPCR/G Protein and neuronal signaling pathways
  • Long-acting, stops signaling between neurons and leads to muscle relaxation
  • Used in the management of chronic obstructive pulmonary disease
  • Improves symptoms of chronic obstructive pulmonary disease complicated by chronic heart failure
Catalog No. 50-003-85179 Supplier Medchemexpress LLC Supplier No. HY1736010MM/1ML
Tiotropium bromide is a muscarinic acetylcholine receptor (mAChR) antagonist that blocks the binding of the acetylcholine ligand and subsequent opening of the ligand-gated ion channel. It is a 24-hour, anticholinergic bronchodilator used in the management of chronic obstructive pulmonary disease. It improves symptoms of chronic obstructive pulmonary disease complicated by chronic heart failure.
  • Targets mAChR
  • Involved in GPCR/G Protein and neuronal signaling pathways
  • Long-acting, stops signaling between neurons and leads to muscle relaxation
  • Used in the management of chronic obstructive pulmonary disease
  • Improves symptoms of chronic obstructive pulmonary disease complicated by chronic heart failure