Learn More
Medchemexpress LLC Tiotropium (Bromide) | 136310-93-5 | 99.9% | 472.42 | 100 MG
SDP
Catalog No. 5000385676
Encompass_Preferred
Tiotropium Bromide is a muscarinic acetylcholine receptor (mAChR) antagonist that blocks the binding of the acetylcholine ligand and the subsequent opening of the ligand-gated ion channel. It is a long-acting, 24-hour, anticholinergic bronchodilator used in the management of chronic obstructive pulmonary disease (COPD). It has also been shown to improve symptoms of COPD complicated by chronic heart failure.
  • Muscarinic acetylcholine receptor (mAChR) antagonist
  • Blocks binding of acetylcholine ligand and opening of ligand-gated ion channel
  • Stops signaling between neurons leading to muscle relaxation
  • Long-acting, 24-hour anticholinergic bronchodilator
  • Used in the management of chronic obstructive pulmonary disease (COPD)
  • Improves symptoms of COPD complicated by chronic heart failure
  • For research use only
Catalog No. 50-003-85676 Supplier Medchemexpress LLC Supplier No. HY17360100MG
Tiotropium Bromide is a muscarinic acetylcholine receptor (mAChR) antagonist that blocks the binding of the acetylcholine ligand and the subsequent opening of the ligand-gated ion channel. It is a long-acting, 24-hour, anticholinergic bronchodilator used in the management of chronic obstructive pulmonary disease (COPD). It has also been shown to improve symptoms of COPD complicated by chronic heart failure.
  • Muscarinic acetylcholine receptor (mAChR) antagonist
  • Blocks binding of acetylcholine ligand and opening of ligand-gated ion channel
  • Stops signaling between neurons leading to muscle relaxation
  • Long-acting, 24-hour anticholinergic bronchodilator
  • Used in the management of chronic obstructive pulmonary disease (COPD)
  • Improves symptoms of COPD complicated by chronic heart failure
  • For research use only