Learn More
Medchemexpress LLC Tiotropium Bromide | 136310-93-5 | 99.9% | 472.42 | 5 MG
SDP
Catalog No. 5000385989
Encompass_Preferred
Tiotropium Bromide is a muscarinic acetylcholine receptor (mAChR) antagonist that blocks the binding of the acetylcholine ligand and the subsequent opening of the ligand-gated ion channel. This compound stops signaling between neurons, leading to muscle relaxation. It is also a long-acting, 24-hour, anticholinergic bronchodilator used in the management of chronic obstructive pulmonary disease and can improve symptoms of chronic obstructive pulmonary disease complicated by chronic heart failure.
  • mAChR antagonist
  • Blocks binding of acetylcholine ligand
  • Blocks subsequent opening of ligand-gated ion channel
  • Long-acting, 24-hour anticholinergic bronchodilator
  • Stops signaling between neurons
  • Leads to muscle relaxation
  • Used in the management of chronic obstructive pulmonary disease (COPD)
  • Improves symptoms of COPD complicated by chronic heart failure
Catalog No. 50-003-85989 Supplier Medchemexpress LLC Supplier No. HY173605MG
Tiotropium Bromide is a muscarinic acetylcholine receptor (mAChR) antagonist that blocks the binding of the acetylcholine ligand and the subsequent opening of the ligand-gated ion channel. This compound stops signaling between neurons, leading to muscle relaxation. It is also a long-acting, 24-hour, anticholinergic bronchodilator used in the management of chronic obstructive pulmonary disease and can improve symptoms of chronic obstructive pulmonary disease complicated by chronic heart failure.
  • mAChR antagonist
  • Blocks binding of acetylcholine ligand
  • Blocks subsequent opening of ligand-gated ion channel
  • Long-acting, 24-hour anticholinergic bronchodilator
  • Stops signaling between neurons
  • Leads to muscle relaxation
  • Used in the management of chronic obstructive pulmonary disease (COPD)
  • Improves symptoms of COPD complicated by chronic heart failure