Learn More
Medchemexpress LLC Tiotropium Bromide | 136310-93-5 | 99.9% | 472.42 | 500 MG
SDP
Catalog No. 5000384960
Encompass_Preferred
Tiotropium Bromide is a potent muscarinic acetylcholine receptor (mAChR) antagonist. It works by blocking the binding of acetylcholine, which prevents the opening of ligand-gated ion channels. This action leads to the cessation of neuronal signaling and muscle relaxation. As a long-acting, 24-hour anticholinergic bronchodilator, it is widely used in the management of chronic obstructive pulmonary disease and can also alleviate symptoms in cases complicated by chronic heart failure.
  • mAChR antagonist
  • Blocks acetylcholine binding
  • Leads to muscle relaxation
  • 24-hour anticholinergic bronchodilator
  • Manages chronic obstructive pulmonary disease
  • Improves symptoms in chronic heart failure complications
Catalog No. 50-003-84960 Supplier Medchemexpress LLC Supplier No. HY17360500MG
Tiotropium Bromide is a potent muscarinic acetylcholine receptor (mAChR) antagonist. It works by blocking the binding of acetylcholine, which prevents the opening of ligand-gated ion channels. This action leads to the cessation of neuronal signaling and muscle relaxation. As a long-acting, 24-hour anticholinergic bronchodilator, it is widely used in the management of chronic obstructive pulmonary disease and can also alleviate symptoms in cases complicated by chronic heart failure.
  • mAChR antagonist
  • Blocks acetylcholine binding
  • Leads to muscle relaxation
  • 24-hour anticholinergic bronchodilator
  • Manages chronic obstructive pulmonary disease
  • Improves symptoms in chronic heart failure complications