Learn More
Apexbio Technology LLC Tiotropium Bromide 136310-93-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064002
Encompass_Preferred
Tiotropium Bromide (CAS 136310-93-5) is a potent antagonist of muscarinic acetylcholine receptors (mAChRs) By competitively inhibiting acetylcholine binding at these receptors it prevents receptor activation and subsequent signaling cascades leading to decreased neuronal transmission and smooth muscle relaxation As a long-acting anticholinergic bronchodilator with activity lasting approximately 24 hours Tiotropium Bromide is widely studied in managing chronic obstructive pulmonary disease (COPD) including cases complicated by chronic heart failure
Catalog No. 50-000-64002 Supplier Apexbio Technology LLC Supplier No. A38745.1
Tiotropium Bromide (CAS 136310-93-5) is a potent antagonist of muscarinic acetylcholine receptors (mAChRs) By competitively inhibiting acetylcholine binding at these receptors it prevents receptor activation and subsequent signaling cascades leading to decreased neuronal transmission and smooth muscle relaxation As a long-acting anticholinergic bronchodilator with activity lasting approximately 24 hours Tiotropium Bromide is widely studied in managing chronic obstructive pulmonary disease (COPD) including cases complicated by chronic heart failure