Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tiotropium Bromide Monohydrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014159 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4159 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4159 Supplier MilliporeSigma Supelco Supplier No. PHR189050MG
Add to Cart
Add to Cart

Tiotropium bromide monohydrate is structurally related to ipratropium bromide. It is a second-generation anticholinergic agent specific for muscarinic receptors (M1, M2 & M3) in humans.

TRUSTED_SUSTAINABILITY

Specifications

CAS 411207-31-3
Linear Formula C19H22 BrNO4S2 · H2O
MDL Number MFCD23704176
Synonym Tiotropium bromide monohydrate
Type Certified Reference Material
Quantity 50 mg
Formula Weight 490.43
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.